Avidin
Proveedor: G Biosciences
786-581 786-582 786-583
786-581EA 137 EUR
786-581 786-582 786-583
Avidin
Proteínas y Péptidos
Avidin is a glycoprotein with approximately 10% of its total mass comes from carbohydrates. Avidin has a molecular weight of 67 kDa and contains four identical 128 amino acid subunits that each have a single biotin binding domain. This affinity purified avidin is ideal for estimation of biotin incorporation and other applications.