Sterile swabs, with PE heads
Proveedor: Texwipe
STX763 STX764
115-1885EA 107 EUR
115-1885 115-1886
Sterile swabs, with PE heads
Hisopos Hisopos para limpieza
Sterile swabs are constructed from 100% medical grade spun polyester. These swabs are bonded securely to a PP shaft by an aqueous-based adhesive.
  • Lot number and expiration date clearly marked and included in barcode for inventory control
  • Gamma irradiated to eliminate repackaging, documentation, and validation
  • Visible through clear packaging to offers visibility of seal integrity
Made to exacting and consistent tolerances using high precision automated processes.
Certificaciones: Certificates of irradiation and compliance attached to each caseMore Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Longitud de cabezal 18 mm 17 mm
Ancho de cabezal 5,8 mm 4,6 mm
Longitud de asa 137 mm 137 mm
Ancho de asa 2,5 mm 2,5 mm