HPLC capillary columns, ZORBAX
Proveedor: Agilent
5064-8253 5064-8254 5064-8255 5064-8256 5064-8257 5064-8258 5064-8260 5064-8261 5064-8263 5064-8264 5064-8265 5064-8266 5064-8267 5064-8269 5064-8271 5064-8286 5064-8287 5064-8291 5064-8294 5064-8295 5064-8296 5064-8297 5064-8298 5064-8270 5065-4459 5065-4460 5065-4462 5065-4464 5065-4466 5064-8300 5064-8259 5065-4461 5065-4463 5065-4465 5065-4467 5065-4468 5065-9912 5065-9942
HEWL5064-8253EA 0 EUR
HEWL5064-8253 HEWL5064-8254 HEWL5064-8255 HEWL5064-8256 HEWL5064-8257 HEWL5064-8258 HEWL5064-8260 HEWL5064-8261 HEWL5064-8263 HEWL5064-8264 HEWL5064-8265 HEWL5064-8266 HEWL5064-8267 HEWL5064-8269 HEWL5064-8271 HEWL5064-8286 HEWL5064-8287 HEWL5064-8291 HEWL5064-8294 HEWL5064-8295 HEWL5064-8296 HEWL5064-8297 HEWL5064-8298 HEWL5064-8270 HEWL5065-4459 HEWL5065-4460 HEWL5065-4462 HEWL5065-4464 HEWL5065-4466 HEWL5064-8300 HEWL5064-8259 HEWL5065-4461 HEWL5065-4463 HEWL5065-4465 HEWL5065-4467 HEWL5065-4468 HEWL5065-9912 HEWL5065-9942
HPLC capillary columns, ZORBAX
Columnas de cromatografía
Columns are ideal for very sample-limited applications because they provide enhanced sensitivity by reducing on-column sample dilution. This high sensitivity can be provided with exceptional reproducibility using columns and low dispersion HPLC instruments.
  • Highest sensitivity for all smallest sample sizes
  • Compatible with all LC/MS interfaces
  • I.Ø of 0,5, 0,3, 0,1, and 0,075 mm
  • Packings/phases for both small and large molecules (80 and 300 Å pore sizes, respectively)
  • Ideal for 1-D and 2-D (proteomics) applications