Heating thermostats, Proline
Proveedor: LAUDA
L001496 L001497 L001498 L001499 L001500 L001501 L001502 L001503 L001504 L001505 L001506 L001507 L001508 L001509 L001489
LAUDL001496EA 3890 EUR
LAUDL001496 LAUDL001497 LAUDL001498 LAUDL001499 LAUDL001500 LAUDL001501 LAUDL001502 LAUDL001503 LAUDL001504 LAUDL001505 LAUDL001506 LAUDL001507 LAUDL001508 LAUDL001509 461-1219
Heating thermostats, Proline
Termostatos con circulación Sistemas de circulación abierta Baños con circuladores integrales
The Proline Edition X range of heating thermostats feature a wide temperature range, high cooling and heating power and a multi level pump. The self-explanatory menu-guided remote control Command is particularly convenient for operating the units at distances as far as 50 meters away. Thanks to the large display, the user can keep an eye on all the important parameters. There is a graphical display of measured values, large choice of interfaces (e.g. Ether- CAT, Ethernet, RS232/RS485, analogue module, Profibus, contact modules). Data collection together with documentation and control is easy using the Wintherm Plus software. Units are ideal for temperature control of chemical reactors, distillation plants and glass condensers, crystallisation, thermostating measurement setups in process technology, chemical probes, calibration of sensors and testing electronic components.
  • Easy to use with simple, intuitive control
  • PowerAdapt system provides maximum heating performance, up to 3,5 kW depending on the network in use
  • SmartCool system gives to 75% energy savings thanks to digital refrigeration management
  • Powerful Varioflex pump (pressure-suction pump) with eight pump levels (except model P12 C)
  • Flexible system with various interface modules, lateral and rear pump connections
  • Multifunctional: Simultaneous connection of two external applications is possible
Max. flow rate / max. pressure:
25 L/min pressure and 23 L/min suction (except P 12 C: 32 L/min pressure only)
0,7 bar pump pressure and 0,4 bar suction performance (except P 12 C: 1.1 bar pressure only)
Certificaciones: IP protection class according to IEC529; IP 21 (except P 40 C and P 50 C: IP 20)
Información de suministro: Supplied with Wintherm Plus software, a wide selection of accessories are available these must be ordered separately.