Amino acid F for Rapid liquid silica method
Proveedor: Hach
2353011
2651155
HACH2353011EA
41.8
EUR
HACH2353011
HACH2651155
Amino acid F for Rapid liquid silica method
Kits para test medioambiente
These are used for determination of Ultra Low Range silica by the Heteropoly Blue Rapid Liquid method. Rapid Liquid™ methods offer an economical way to analyse multiple samples and to measure trace concentrations.