Capillary columns, GS-CarbonPLOT
Proveedor: Agilent
113-3112 113-3132 113-3133 113-3162 115-3113 115-3133 113-3133LTM 115-3133LTM
HEWL113-3112EA 528 EUR
HEWL113-3112 HEWL113-3132 HEWL113-3133 HEWL113-3162 HEWL115-3113 HEWL115-3133 HEWL113-3133LTM HEWL115-3133LTM
Capillary columns, GS-CarbonPLOT
Columnas de cromatografía
These columns are ideal for GC/MS.
  • High stability, carbon layer stationary phase
  • Unique selectivity for inorganic and organic gases
  • Extended temperature limit of 360 ºC
  • Retention stability not affected by water
Bonded monolithic carbon layer.
Similar phases: Carbopack, CLOT, Carboxen-1006 PLOT.