Capillary columns, HP-5ms Ultra Inert
Proveedor: Agilent
19091S-133UI 19091S-213UI 19091S-233UI 19091S-413UI 19091S-431UI 19091S-433UI 19091S-433UIE 19091S-436UI 19091S-577UI
HEWL19091S-133UIEA 901 EUR
HEWL19091S-133UI HEWL19091S-213UI HEWL19091S-233UI HEWL19091S-413UI HEWL19091S-431UI HEWL19091S-433UI HEWL19091S-433UIE HEWL19091S-436UI HEWL19091S-577UI
Capillary columns, HP-5ms Ultra Inert
Columnas de cromatografía
These columns are recommended for semivolatiles, halogenated compounds, pesticides, herbicides, drugs of abuse, amines, unknown sample screening.
  • Excellent for trace level analysis of acids and bases, such as semivolatiles analysis
  • Specifically tested for inertness for active compounds including acidic and basic compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Non-polar with very low bleed characteristics, ideal for GC/MS
  • Bonded, cross-linked and solvent rinsable
5% Phenyl 95% dimethylpolysiloxane