Puregene RBC and Cell Lysis Solutions
Proveedor: QIAGEN
158904
158908
QIAG158904EA
0
EUR
QIAG158904
QIAG158908
Puregene RBC and Cell Lysis Solutions
Tampones
Tampones de extracción
For purification of archive-quality DNA from a wide range of sample types.
- Archive-quality DNA for long-term storage
- Reproducible DNA purification
- Convenient, scalable purification procedure
Puregene Accessories used in combination with Puregene Kits enable purification of high-molecular-weight (100–200 kb) DNA suitable for archiving. The scalable purification procedure gently removes contaminants and inhibitors and allows large-volume samples to be purified for use as long-term references.