Anti-MLKL Rabbit Polyclonal Antibody

Proveedor: Abgent
ABGEAP14272BEA 348 EUR
ABGEAP14272B
Anti-MLKL Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MLKL (mixed lineage kinase domain-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR