Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5]

Proveedor: Tonbo Biosciences
40-0041-U500 40-0041-M001 40-0041-U100
40-0041-U500EA 147 EUR
40-0041-U500 40-0041-M001 40-0041-U100
Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5]
Anticuerpos
The GK1.5 antibody reacts with mouse CD4, a 55 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4 is typically expressed on thymocytes, certain mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells

The GK1,5 antibody is widely used as a phenotypic marker for CD4 expression. If used together, the GK1,5 antibody and an alternative antibody, Anti-Mouse CD4 clone RM4-5, will 'compete' for binding, i.e. RM4-5 is able to block GK1,5 binding to cells. In contrast, the Anti-Mouse CD4 clone RM4-4 does not block binding of the GK1,5 antibody to cells (Arora S et al. 2006. Infect. Immun. 74: 4339-4348). The GK1,5 antibody is also reported to be cross-reactive with Syrian hamster CD4.

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: GK1.5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR