Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RM4-5]

Proveedor: Tonbo Biosciences
60-0042-U100 60-0042-U025
60-0042-U100EA 129 EUR
60-0042-U100 60-0042-U025
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RM4-5]
Anticuerpos
The RM4-5 antibody reacts with mouse CD4, a 55 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta2-domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4 is typically expressed on thymocytes, certain mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.

The RM4-5 antibody is widely used as a phenotypic marker for CD4 expression. If used together, the RM4-5 antibody and an alternative antibody, Anti-Mouse CD4 clone GK1.5, will “compete” for binding, i.e. RM4-5 antibody is able to block GK1.5 antibody binding to cells. In contrast, RM4-5 antibody does not block the binding of Anti-Mouse CD4 clone RM4-4 to cells.

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: RM4-5
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR