Anti-CD45RA Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: HI100]

Proveedor: Tonbo Biosciences
65-0458-T100 65-0458-T025
65-0458-T100EA 168 EUR
65-0458-T100 65-0458-T025
Anti-CD45RA Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: HI100]
Anticuerpos
The HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RB and CD45RO.

The HI100 antibody is widely used as a marker for human CD45RA expression on naïve and activated T cells, B cells, and monocytes.

Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: monoclonal
Clone: HI100
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR