Anti-CD19 Mouse Monoclonal Antibody (violetFluor® 450) [clone: HIB19]

Proveedor: Tonbo Biosciences
75-0199-T100 75-0199-T025
75-0199-T100EA 225 EUR
75-0199-T100 75-0199-T025
Anti-CD19 Mouse Monoclonal Antibody (violetFluor® 450) [clone: HIB19]
Anticuerpos
The HIB19 antibody reacts with human CD19, a 95 kDa glycoprotein which acts as a co-receptor, along with CD21, CD81 and CD225, in support of the functional B cell receptor (BCR). This complex provides antigen-specific recognition and subsequent activation of B cells to proliferate and differentiate into antibody-secreting cells (plasma cells) or memory B cells, which are crucial for secondary antigen encounter. CD19 is a lineage-differentiation marker, as its expression is detectable at the earliest B cell stages, through development, and is finally lost upon transition to mature plasma cells.

The HIB19 antibody is widely used as a phenotypic marker for CD19 expression on B cells, as well as on dendritic cell subsets.

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: HIB19
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR