Anti-CACNA1S Rabbit polyclonal antibody

Proveedor: Biorbyt
ORB1137-100 ORB1137-200
BIRBORB1137-100EA 0 EUR
BIRBORB1137-100 BIRBORB1137-200
Anti-CACNA1S Rabbit polyclonal antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CACNA1S (calcium channel, voltage-dependent, L type, alpha 1S subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR