Anti-ENAH Rabbit polyclonal antibody

Proveedor: Biorbyt
ORB1367-100 ORB1367-200
BIRBORB1367-100EA 0 EUR
BIRBORB1367-100 BIRBORB1367-200
Anti-ENAH Rabbit polyclonal antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: ENAH (enabled homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR