Anti-C2orf43 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB1418-100 ORB1418-200
BIRBORB1418-100EA 0 EUR
BIRBORB1418-100 BIRBORB1418-200
Anti-C2orf43 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: C2orf43 (chromosome 2 open reading frame 43)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR