Anti-C3orf18 Rabbit polyclonal antibody
Proveedor: Biorbyt
ORB1419-100 ORB1419-200
BIRBORB1419-100EA 0 EUR
BIRBORB1419-100 BIRBORB1419-200
Anti-C3orf18 Rabbit polyclonal antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: C3orf18 (chromosome 3 open reading frame 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now