Anti-PROCA1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB1703-100 ORB1703-200
BIRBORB1703-100EA 0 EUR
BIRBORB1703-100 BIRBORB1703-200
Anti-PROCA1 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: PROCA1 (protein interacting with cyclin A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR