Anti-MROH2B Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB2133-100 ORB2133-200
BIRBORB2133-100EA 0 EUR
BIRBORB2133-100 BIRBORB2133-200
Anti-MROH2B Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: MROH2B (maestro heat-like repeat family member 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR