Anti-FSIP1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB2964-100 ORB2964-200
BIRBORB2964-100EA 0 EUR
BIRBORB2964-100 BIRBORB2964-200
Anti-FSIP1 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: FSIP1 (fibrous sheath interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR