Anti-SIGLEC11 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
ORB4868-100 ORB4868-200
BIRBORB4868-100EA 0 EUR
BIRBORB4868-100 BIRBORB4868-200
Anti-SIGLEC11 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: SIGLEC11 (sialic acid binding Ig-like lectin 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat