Anti-Chloramphenicol Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB10383-100 ORB10383-200
BIRBORB10383-100EA 0 EUR
BIRBORB10383-100 BIRBORB10383-200
Anti-Chloramphenicol Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000 (based on 0.5 mg/ml)

Type: Primary
Antigen: Chloramphenicol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR