Anti-HBcAg Mouse monoclonal antibody

Proveedor: Biorbyt
ORB10775-100 ORB10775-200
BIRBORB10775-100EA 0 EUR
BIRBORB10775-100 BIRBORB10775-200
Anti-HBcAg Mouse monoclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HBcAg
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR