Anti-SR-B1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
ORB10791-100 ORB10791-200
BIRBORB10791-100EA 0 EUR
BIRBORB10791-100 BIRBORB10791-200
Anti-SR-B1 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: SR-B1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now