Anti-HIST3H3 Rabbit polyclonal antibody

Proveedor: Biorbyt
ORB10805-100 ORB10805-200
BIRBORB10805-100EA 0 EUR
BIRBORB10805-100 BIRBORB10805-200
Anti-HIST3H3 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR