Anti-NFKB1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
ORB11124-100 ORB11124-200
BIRBORB11124-100EA 0 EUR
BIRBORB11124-100 BIRBORB11124-200
Anti-NFKB1 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: NFKB1 (nuclear factor of kappa light polypeptide gene enhancer in B-cells 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now