Anti-CD79A Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB11330-100 ORB11330-200
BIRBORB11330-100EA 0 EUR
BIRBORB11330-100 BIRBORB11330-200
Anti-CD79A Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Secondary
Antigen: CD79A (CD79a molecule, immunoglobulin-associated alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR