Anti-Shiga-like Toxin 2 (SLT2) Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB11367-100 ORB11367-20 ORB11367-200 ORB11367-5
BIRBORB11367-100EA 0 EUR
BIRBORB11367-100 BIRBORB11367-20 BIRBORB11367-200 BIRBORB11367-5
Anti-Shiga-like Toxin 2 (SLT2) Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000

Type: Primary
Antigen: Shiga-like Toxin 2 (SLT2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR