Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin))

Proveedor: Biorbyt
ORB12064-100 ORB12064-50
BIRBORB12064-100EA 0 EUR
BIRBORB12064-100 BIRBORB12064-50
Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR