Anti-PTPRC Mouse Monoclonal Antibody (Biotin)

Proveedor: Biorbyt
ORB12066-100 ORB12066-500
BIRBORB12066-100EA 0 EUR
BIRBORB12066-100 BIRBORB12066-500
Anti-PTPRC Mouse Monoclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR