Anti-CD59 Mouse Monoclonal Antibody

Proveedor: Biorbyt
ORB12103-100 ORB12103-500
BIRBORB12103-100EA 0 EUR
BIRBORB12103-100 BIRBORB12103-500
Anti-CD59 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR