Anti-BSG Mouse Monoclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
ORB12139-100 ORB12139-50
BIRBORB12139-100EA 0 EUR
BIRBORB12139-100 BIRBORB12139-50
Anti-BSG Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR