Anti-LH/CG Receptor Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB13542-100 ORB13542-200
BIRBORB13542-100EA 0 EUR
BIRBORB13542-100 BIRBORB13542-200
Anti-LH/CG Receptor Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: LH/CG Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR