Anti-ORF73 Mouse Monoclonal Antibody

Proveedor: Biorbyt
ORB22725-100 ORB22725-20 ORB22725-5
BIRBORB22725-100EA 0 EUR
BIRBORB22725-100 BIRBORB22725-20 BIRBORB22725-5
Anti-ORF73 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: ORF73
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR