Anti-PSMA3 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB49148-100 ORB49148-200
BIRBORB49148-100EA 0 EUR
BIRBORB49148-100 BIRBORB49148-200
Anti-PSMA3 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PSMA3 (proteasome (prosome, macropain) subunit, alpha type, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR