Anti-TNIP3 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB100196-100 ORB100196-200
BIRBORB100196-100EA 0 EUR
BIRBORB100196-100 BIRBORB100196-200
Anti-TNIP3 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: TNIP3 (TNFAIP3 interacting protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR