Anti-UTS2B Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB100386-100 ORB100386-200
BIRBORB100386-100EA 0 EUR
BIRBORB100386-100 BIRBORB100386-200
Anti-UTS2B Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: UTS2B (urotensin 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR