Anti-Staphyloccocus aureus Rosenbach tropina Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB100467-100 ORB100467-200
BIRBORB100467-100EA 0 EUR
BIRBORB100467-100 BIRBORB100467-200
Anti-Staphyloccocus aureus Rosenbach tropina Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: Staphyloccocus aureus Rosenbach tropina
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR