Anti-BNIP2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB100558-100 ORB100558-200
BIRBORB100558-100EA 0 EUR
BIRBORB100558-100 BIRBORB100558-200
Anti-BNIP2 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: BNIP2 (BCL2/adenovirus E1B 19kDa interacting protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR