Anti-4-Hydroxynonenal Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB100588-100 ORB100588-200
BIRBORB100588-100EA 0 EUR
BIRBORB100588-100 BIRBORB100588-200
Anti-4-Hydroxynonenal Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: 4-Hydroxynonenal (4-hydroxynonenal)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR