Anti-FASTKD1 Rabbit polyclonal antibody

Proveedor: Biorbyt
ORB100798-100 ORB100798-200
BIRBORB100798-100EA 0 EUR
BIRBORB100798-100 BIRBORB100798-200
Anti-FASTKD1 Rabbit polyclonal antibody
Anticuerpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: FASTKD1 (FAST kinase domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR