Anti-Coronin 3 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB101746-100 ORB101746-200
BIRBORB101746-100EA 0 EUR
BIRBORB101746-100 BIRBORB101746-200
Anti-Coronin 3 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: Coronin 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR