Anti-IRS1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
ORB106419-100 ORB106419-20
BIRBORB106419-100EA 0 EUR
BIRBORB106419-100 BIRBORB106419-20
Anti-IRS1 Rabbit Polyclonal Antibody
Anticuerpos
Immunochemistry: 1:50-1:200
Type: Primary
Antigen: IRS1 (insulin receptor substrate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat