Anti-AKIP1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB125481-100 ORB125481-200
BIRBORB125481-100EA 0 EUR
BIRBORB125481-100 BIRBORB125481-200
Anti-AKIP1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: AKIP1 (A kinase (PRKA) interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR