Anti-NOS2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB126045-100 ORB126045-200
BIRBORB126045-100EA 0 EUR
BIRBORB126045-100 BIRBORB126045-200
Anti-NOS2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR