Anti-PEBP1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
ORB126235-100 ORB126235-200
BIRBORB126235-100EA 0 EUR
BIRBORB126235-100 BIRBORB126235-200
Anti-PEBP1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PEBP1 (phosphatidylethanolamine binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human