Anti-MLTK Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB127068-100 ORB127068-200
BIRBORB127068-100EA 0 EUR
BIRBORB127068-100 BIRBORB127068-200
Anti-MLTK Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MLTK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR