Anti-TAF13 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB127259-100 ORB127259-200
BIRBORB127259-100EA 0 EUR
BIRBORB127259-100 BIRBORB127259-200
Anti-TAF13 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TAF13 (TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 18kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR