Anti-GAB2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB127367-100 ORB127367-200
BIRBORB127367-100EA 0 EUR
BIRBORB127367-100 BIRBORB127367-200
Anti-GAB2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: GAB2 (GRB2-associated binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR