Anti-PRKCD Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB127468-100 ORB127468-200
BIRBORB127468-100EA 0 EUR
BIRBORB127468-100 BIRBORB127468-200
Anti-PRKCD Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PRKCD (protein kinase C, delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR